Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130417.2 | 2773 | UTR 3 | NP_001123889.1 | ||
NM_033262.4 | 2773 | UTR 3 | NP_150287.1 | ||
NM_058240.3 | 2773 | UTR 3 | NP_489479.1 | ||
NM_182932.2 | 2773 | UTR 3 | NP_891977.1 | ||
NM_182936.2 | 2773 | UTR 3 | NP_891981.1 | ||
NM_183002.2 | 2773 | UTR 3 | NP_892114.1 | ||
XM_006720240.3 | 2773 | UTR 3 | XP_006720303.1 | ||
XM_017021606.1 | 2773 | UTR 3 | XP_016877095.1 | ||
XM_017021607.1 | 2773 | UTR 3 | XP_016877096.1 | ||
XM_017021608.1 | 2773 | UTR 3 | XP_016877097.1 | ||
XM_017021609.1 | 2773 | UTR 3 | XP_016877098.1 | ||
XM_017021610.1 | 2773 | Intron | XP_016877099.1 | ||
XM_017021611.1 | 2773 | UTR 3 | XP_016877100.1 |