Product Details

SNP ID
rs344954
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:188178943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGCCTCTGTGGACATGCAGGTAT[A/G]TTAGGCCAGAGGTGGGGAGTGGGAG
Phenotype
MIM: 600700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ42393 PubMed Links

Gene Details

Gene
FLJ42393
Gene Name
uncharacterized LOC401105
There are no transcripts associated with this gene.

Gene
LPP
Gene Name
LIM domain containing preferred translocation partner in lipoma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167671.2 Intron NP_001161143.1
NM_001167672.2 Intron NP_001161144.1
NM_005578.4 Intron NP_005569.1
XM_005247446.4 Intron XP_005247503.1
XM_005247450.4 Intron XP_005247507.1
XM_005247451.4 Intron XP_005247508.1
XM_005247453.2 Intron XP_005247510.1
XM_011512820.2 Intron XP_011511122.1
XM_011512823.2 Intron XP_011511125.1
XM_011512827.2 Intron XP_011511129.1
XM_011512828.2 Intron XP_011511130.1
XM_011512831.2 Intron XP_011511133.1
XM_011512833.2 Intron XP_011511135.1
XM_011512834.2 Intron XP_011511136.1
XM_011512836.2 Intron XP_011511138.1
XM_017006377.1 Intron XP_016861866.1
XM_017006378.1 Intron XP_016861867.1
XM_017006379.1 Intron XP_016861868.1
XM_017006380.1 Intron XP_016861869.1
XM_017006381.1 Intron XP_016861870.1
XM_017006382.1 Intron XP_016861871.1

View Full Product Details