Product Details

SNP ID
rs495078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45829910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCCCACCCCAGCAGACCTAATT[C/T]TTGTTAAAGTGCCATTCTGTAAGGG
Phenotype
MIM: 615068
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPG5 PubMed Links

Gene Details

Gene
EPG5
Gene Name
ectopic P-granules autophagy protein 5 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020964.2 Intron NP_066015.2
XM_011526122.1 Intron XP_011524424.1
XM_017025889.1 Intron XP_016881378.1
XM_017025890.1 Intron XP_016881379.1
XM_017025891.1 Intron XP_016881380.1
XM_017025892.1 Intron XP_016881381.1
XM_017025893.1 Intron XP_016881382.1
Gene
SIGLEC15
Gene Name
sialic acid binding Ig like lectin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213602.2 Intron NP_998767.1

View Full Product Details