Product Details

SNP ID
rs631982
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:62436341 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGCCTCGCTCCGGAAGCCGCAC[C/T]CTCCGCGGCGTCCCACCGTCGAGGA
Phenotype
MIM: 603478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP1 PubMed Links

Gene Details

Gene
USP1
Gene Name
ubiquitin specific peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017415.1 38 UTR 5 NP_001017415.1
NM_001017416.1 38 UTR 5 NP_001017416.1
NM_003368.4 38 Intron NP_003359.3

View Full Product Details