Product Details
- SNP ID
-
rs745182
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:80141966 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACTGACCCCCACCTGGGCCGTCCTC[C/T]GCTTGCTCCTGAGTTTGCTTTGAGG
- Phenotype
-
MIM: 612857
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PLAC9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114929421] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLAC9
- Gene Name
- placenta specific 9
View Full Product Details