Product Details
- SNP ID
-
rs380955
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:158751100 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATTCCATTATTACAGTTCTATTT[C/T]AGGCCTTTACTCCAGATTATTGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6K6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111372019] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6K6
- Gene Name
- olfactory receptor family 6 subfamily K member 6
There are no transcripts associated with this gene.
- Gene
- OR6N1
- Gene Name
- olfactory receptor family 6 subfamily N member 1
View Full Product Details