Product Details

SNP ID
hCV899625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127419950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCAGTGCCTCCAGAATGTGGC[A/C]GCTCACAAGCCTCCTGCTGTTCGTG
Phenotype
MIM: 612283
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR4783 PubMed Links

Gene Details

Gene
MIR4783
Gene Name
microRNA 4783
There are no transcripts associated with this gene.

Gene
PROC
Gene Name
protein C, inactivator of coagulation factors Va and VIIIa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000312.3 102 Missense Mutation CAG,CCG Q3P NP_000303.1

View Full Product Details