Product Details

SNP ID
rs260218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110051095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACTCTGCTGGAGAGTTCATTTC[A/G]CTTTCTTTCTGGGATGCTGAGATGC
Phenotype
MIM: 604582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKAP2 PubMed Links

Gene Details

Gene
AKAP2
Gene Name
A-kinase anchoring protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004065.4 Intron NP_001004065.2
NM_001136562.2 Intron NP_001130034.1
NM_001198656.1 Intron NP_001185585.1
Gene
PALM2-AKAP2
Gene Name
PALM2-AKAP2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007203.4 Intron NP_009134.1
NM_147150.2 Intron NP_671492.1

View Full Product Details