Product Details
- SNP ID
-
rs758585
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:114822801 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCGACAGCTTCCAAGCTCACCGAGC[A/C]CAGAGACCATCTGTTTCCTTTCTTG
- Phenotype
-
MIM: 602330
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABLIM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76905475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABLIM1
- Gene Name
- actin binding LIM protein 1
There are no transcripts associated with this gene.
- Gene
- FAM160B1
- Gene Name
- family with sequence similarity 160 member B1
View Full Product Details