Product Details

SNP ID
rs747830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80628700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATATTCATTGTTACAAGCATTA[C/T]AGAACAGAAAAGTCCAAGGACCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF704 PubMed Links

Gene Details

Gene
ZNF704
Gene Name
zinc finger protein 704
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033723.2 14768 UTR 3 NP_001028895.1
XM_005251279.4 14768 UTR 3 XP_005251336.2
XM_017013725.1 14768 UTR 3 XP_016869214.1

View Full Product Details