Product Details

SNP ID
rs12806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100151385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCAACCCAATGCAGGGAATGCA[A/G]GGCCCCAGAGCTTTGGCTCAGAGCA
Phenotype
MIM: 611604
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERLIN1 PubMed Links

Gene Details

Gene
ERLIN1
Gene Name
ER lipid raft associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100626.1 1900 UTR 3 NP_001094096.1
NM_006459.3 1900 UTR 3 NP_006450.2
XM_005269442.3 1900 UTR 3 XP_005269499.1
XM_017015541.1 1900 UTR 3 XP_016871030.1

View Full Product Details