Product Details

SNP ID
rs284249
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:10640748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCCTCTGCCCTCTGCTCAGAG[C/T]CGTAATTTACCTGGGTCAGGACCAG
Phenotype
MIM: 609895 MIM: 601791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASZ1 PubMed Links

Gene Details

Gene
CASZ1
Gene Name
castor zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079843.2 Intron NP_001073312.1
NM_017766.4 Intron NP_060236.3
XM_005263479.3 Intron XP_005263536.1
XM_011541635.2 Intron XP_011539937.1
XM_017001539.1 Intron XP_016857028.1
XM_017001540.1 Intron XP_016857029.1
XM_017001541.1 Intron XP_016857030.1
Gene
PEX14
Gene Name
peroxisomal biogenesis factor 14
There are no transcripts associated with this gene.

View Full Product Details