Product Details

SNP ID
rs506446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31831759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATCCTGTTTGGGGTGTTATAC[A/C]AAAATATTAAGTATACGCTTTATAG
Phenotype
MIM: 614136
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRAPPC8 PubMed Links

Gene Details

Gene
TRAPPC8
Gene Name
trafficking protein particle complex 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014939.3 Intron NP_055754.2
XM_005258233.2 Intron XP_005258290.1
XM_005258234.2 Intron XP_005258291.1
XM_005258235.3 Intron XP_005258292.1
XM_006722420.3 Intron XP_006722483.1
XM_011525884.2 Intron XP_011524186.1
XM_017025615.1 Intron XP_016881104.1
XM_017025616.1 Intron XP_016881105.1
XM_017025617.1 Intron XP_016881106.1

View Full Product Details