Product Details

SNP ID
rs682200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:77542129 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAAGATGGGCTGTAAGAAAAA[A/G]AGCTGGTGAGGGTTGTGTATTAAAA
Phenotype
MIM: 604112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCEL PubMed Links

Gene Details

Gene
SCEL
Gene Name
sciellin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160706.1 Intron NP_001154178.1
NM_003843.3 Intron NP_003834.3
NM_144777.2 Intron NP_659001.2
XM_005266578.1 Intron XP_005266635.1
XM_006719882.1 Intron XP_006719945.1
XM_006719884.1 Intron XP_006719947.1
XM_006719885.1 Intron XP_006719948.1
XM_011535281.1 Intron XP_011533583.1
XM_011535282.1 Intron XP_011533584.1
XM_011535283.1 Intron XP_011533585.1
XM_011535284.1 Intron XP_011533586.1
XM_011535285.1 Intron XP_011533587.1
XM_011535286.1 Intron XP_011533588.1
XM_011535287.1 Intron XP_011533589.1
XM_011535288.1 Intron XP_011533590.1
XM_011535289.1 Intron XP_011533591.1
XM_011535290.1 Intron XP_011533592.1
XM_011535291.1 Intron XP_011533593.1
XM_017020805.1 Intron XP_016876294.1

View Full Product Details