Product Details

SNP ID
rs2019446
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:13371275 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAAATGTTGGTACTATACCATT[C/G]CCTTTCCTGCTCTCCTTTTCTCACT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LINC01559 PubMed Links

Gene Details

Gene
LINC01559
Gene Name
long intergenic non-protein coding RNA 1559
There are no transcripts associated with this gene.

View Full Product Details