Product Details

SNP ID
rs1096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:161289578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGCCATCTATACAAACTTTTACT[A/G]TTTGAAAACTGAGATTTAAGTTGCA
Phenotype
MIM: 600232
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRB2 PubMed Links

Gene Details

Gene
GABRB2
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000813.2 6146 UTR 3 NP_000804.1
NM_021911.2 6146 UTR 3 NP_068711.1

View Full Product Details