Product Details

SNP ID
rs337260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69094376 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCGGAAGGCAGCGACCGGCCGGGT[C/G]GCTCAGGATGCACTTTCTCCGGCCT
Phenotype
MIM: 607819
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC30A5 PubMed Links

Gene Details

Gene
SLC30A5
Gene Name
solute carrier family 30 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251969.1 Intron NP_001238898.1
NM_022902.4 Intron NP_075053.2
NM_024055.4 Intron NP_076960.1
XM_005248569.3 Intron XP_005248626.1
XM_006714672.3 Intron XP_006714735.1
XM_017009749.1 Intron XP_016865238.1

View Full Product Details