Product Details

SNP ID
rs513599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66667689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTGACAAAGGACAATGACCC[A/C]AGAAGTAGGTTTTTTTTTTTTTTTA
Phenotype
MIM: 602571
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBM4 PubMed Links

Gene Details

Gene
RBM4
Gene Name
RNA binding motif protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198843.1 2394 Intron NP_001185772.1
NM_001198844.1 2394 UTR 3 NP_001185773.1
NM_002896.3 2394 Intron NP_002887.2
Gene
RBM4B
Gene Name
RNA binding motif protein 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286135.1 2394 Intron NP_001273064.1
NM_031492.3 2394 Intron NP_113680.1
XM_011545297.2 2394 Intron XP_011543599.1
XM_017018402.1 2394 Intron XP_016873891.1

View Full Product Details