Product Details

SNP ID
rs606967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:124732200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTAACTACTGTAATCGAAGCCTA[C/T]AGTCAGGGCAACTACTCATATTAAT
Phenotype
MIM: 604605 MIM: 613891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KALRN PubMed Links

Gene Details

Gene
KALRN
Gene Name
kalirin, RhoGEF kinase
There are no transcripts associated with this gene.

Gene
MIR544B
Gene Name
microRNA 544b
There are no transcripts associated with this gene.

Gene
UMPS
Gene Name
uridine monophosphate synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000373.3 Intron NP_000364.1

View Full Product Details