Product Details

SNP ID
rs613079
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128698800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGAATTTGGCACTTTACACTA[A/G]AGACACCTCTGGTATTTAATGAAAA
Phenotype
MIM: 193067 MIM: 615815
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FLI1 PubMed Links
Additional Information
For this assay, SNP(s) [rs75977786] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FLI1
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167681.2 Intron NP_001161153.1
NM_001271010.1 Intron NP_001257939.1
NM_001271012.1 Intron NP_001257941.1
NM_002017.4 Intron NP_002008.2
XM_011542701.2 Intron XP_011541003.1
XM_011542702.1 Intron XP_011541004.1
XM_017017405.1 Intron XP_016872894.1
XM_017017406.1 Intron XP_016872895.1
Gene
SENCR
Gene Name
smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA
There are no transcripts associated with this gene.

View Full Product Details