Product Details

SNP ID
rs529502
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:182645168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGCCCCTCACCCCTTAAAAGGA[A/G]AATGGTTCAGCCCAGTTCTCAATCA
Phenotype
MIM: 607189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS8 PubMed Links

Gene Details

Gene
RGS8
Gene Name
regulator of G-protein signaling 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102450.2 2272 UTR 3 NP_001095920.1
NM_033345.3 2272 UTR 3 NP_203131.1
XM_005245555.3 2272 UTR 3 XP_005245612.1
XM_011510089.2 2272 UTR 3 XP_011508391.1
XM_011510090.2 2272 UTR 3 XP_011508392.1
XM_011510091.2 2272 UTR 3 XP_011508393.1
XM_017002631.1 2272 UTR 3 XP_016858120.1
XM_017002632.1 2272 UTR 3 XP_016858121.1
XM_017002633.1 2272 UTR 3 XP_016858122.1
XM_017002634.1 2272 UTR 3 XP_016858123.1
XM_017002635.1 2272 UTR 3 XP_016858124.1
XM_017002636.1 2272 UTR 3 XP_016858125.1
XM_017002637.1 2272 UTR 3 XP_016858126.1

View Full Product Details