Product Details

SNP ID
rs764224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37760995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGTATTCAACCTCAGCACTT[C/T]TGAGGGATATTAAACATTTTCAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMT10B PubMed Links

Gene Details

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 Intron NP_001273879.1
NM_001286951.1 Intron NP_001273880.1
NM_001286952.1 Intron NP_001273881.1
NM_001286953.1 Intron NP_001273882.1
NM_001286954.1 Intron NP_001273883.1
NM_144964.3 Intron NP_659401.2
XM_005251375.2 Intron XP_005251432.1
XM_005251379.2 Intron XP_005251436.1
XM_011517735.2 Intron XP_011516037.1
XM_011517736.2 Intron XP_011516038.1
XM_011517738.2 Intron XP_011516040.1
XM_011517739.2 Intron XP_011516041.1
XM_011517742.1 Intron XP_011516044.1
XM_011517743.2 Intron XP_011516045.1
XM_017014313.1 Intron XP_016869802.1

View Full Product Details