Product Details

SNP ID
rs764226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37760648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATTTCAGTTTAAGGAGTTAATT[A/C]CCAAAAGAACAAAAAAGAACTGGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRMT10B PubMed Links

Gene Details

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 Intron NP_001273879.1
NM_001286951.1 Intron NP_001273880.1
NM_001286952.1 Intron NP_001273881.1
NM_001286953.1 Intron NP_001273882.1
NM_001286954.1 Intron NP_001273883.1
NM_144964.3 Intron NP_659401.2

View Full Product Details