Product Details

SNP ID
rs563751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33584992 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGGTTCAAATGCCAGTTCTAAC[A/G]TGCTGAGCCGTGCGATGGGTGGCAG
Phenotype
MIM: 600516 MIM: 609495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAK1 PubMed Links

Gene Details

Gene
BAK1
Gene Name
BCL2 antagonist/killer 1
There are no transcripts associated with this gene.

Gene
GGNBP1
Gene Name
gametogenetin binding protein 1 (pseudogene)
There are no transcripts associated with this gene.

Gene
LINC00336
Gene Name
long intergenic non-protein coding RNA 336
There are no transcripts associated with this gene.

View Full Product Details