Product Details

SNP ID
rs667690
Assay Type
Functionally Tested
NCBI dbSNP Submissions
28
Location
Chr.1:183252211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTAAAGGGCATGCTGGGAAGGC[A/G]AGAGCCAGAGCCGCCTCCCCAGAGC
Phenotype
MIM: 150292 MIM: 608701
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LAMC2 PubMed Links

Gene Details

Gene
LAMC2
Gene Name
laminin subunit gamma 2
There are no transcripts associated with this gene.

Gene
NMNAT2
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015039.3 1586 UTR 3 NP_055854.1
NM_170706.3 1586 UTR 3 NP_733820.1

View Full Product Details