Product Details

SNP ID
rs273250
Assay Type
Validated
NCBI dbSNP Submissions
64
Location
Chr.1:78649621 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTAAATAAAAAGATTTTTTAAAAGT[C/T]AGTGAGCTGTTATGTTTTCTTCCCT
Phenotype
MIM: 610468 MIM: 613975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI44 PubMed Links

Gene Details

Gene
IFI44
Gene Name
interferon induced protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006417.4 1068 Intron NP_006408.3
XM_005270380.3 1068 UTR 5 XP_005270437.1
XM_011540516.2 1068 UTR 5 XP_011538818.1
XM_017000083.1 1068 UTR 5 XP_016855572.1
Gene
IFI44L
Gene Name
interferon induced protein 44 like
There are no transcripts associated with this gene.

View Full Product Details