Product Details

SNP ID
rs595662
Assay Type
Functionally tested
NCBI dbSNP Submissions
82
Location
Chr.1:70145664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGGCCCAGGCTAATTATACCAAC[A/G]GGTAGGTGATACTGGGAAACTACAA
Phenotype
MIM: 614453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC40 PubMed Links

Gene Details

Gene
LRRC40
Gene Name
leucine rich repeat containing 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017768.4 1825 UTR 3 NP_060238.3
XM_005271013.1 1825 UTR 3 XP_005271070.1
XM_011541763.1 1825 UTR 3 XP_011540065.1
XM_017001695.1 1825 UTR 3 XP_016857184.1
XM_017001696.1 1825 UTR 3 XP_016857185.1
Gene
LRRC7
Gene Name
leucine rich repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020794.2 1825 Intron NP_065845.1
XM_017001885.1 1825 Intron XP_016857374.1
XM_017001886.1 1825 Intron XP_016857375.1
XM_017001887.1 1825 Intron XP_016857376.1
XM_017001888.1 1825 Intron XP_016857377.1
XM_017001889.1 1825 Intron XP_016857378.1
XM_017001890.1 1825 Intron XP_016857379.1
XM_017001891.1 1825 Intron XP_016857380.1
XM_017001892.1 1825 Intron XP_016857381.1
XM_017001893.1 1825 Intron XP_016857382.1
XM_017001894.1 1825 Intron XP_016857383.1
XM_017001895.1 1825 Intron XP_016857384.1

View Full Product Details