Product Details

SNP ID
rs378951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:4498872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAAGAGATATTTACAAGGAGGA[C/G]TAATATTCTGCTCTCCAGCATCCTT
Phenotype
MIM: 147265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITPR1 PubMed Links

Gene Details

Gene
ITPR1
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099952.2 Intron NP_001093422.2
NM_001168272.1 Intron NP_001161744.1
NM_002222.5 Intron NP_002213.5
XM_005265109.2 Intron XP_005265166.1
XM_005265110.2 Intron XP_005265167.1
XM_006713131.2 Intron XP_006713194.1
XM_011533681.1 Intron XP_011531983.1
XM_011533682.2 Intron XP_011531984.1
XM_011533683.2 Intron XP_011531985.1
XM_011533684.1 Intron XP_011531986.1
XM_011533685.1 Intron XP_011531987.1
XM_011533686.1 Intron XP_011531988.1
XM_011533687.1 Intron XP_011531989.1
XM_011533688.1 Intron XP_011531990.1
XM_011533690.1 Intron XP_011531992.1
XM_011533691.1 Intron XP_011531993.1
XM_011533692.2 Intron XP_011531994.1
XM_017006357.1 Intron XP_016861846.1
XM_017006358.1 Intron XP_016861847.1
Gene
ITPR1-AS1
Gene Name
ITPR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details