Product Details

SNP ID
rs32651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:119456137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAATACAGTTTTAAGCTTTCCA[A/G]CCTCTGCAAGCAGAAGAGAGTGGAT
Phenotype
MIM: 601860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD17B4 PubMed Links

Gene Details

Gene
HSD17B4
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000414.3 Intron NP_000405.1
NM_001199291.2 Intron NP_001186220.1
NM_001199292.1 Intron NP_001186221.1
NM_001292027.1 Intron NP_001278956.1
NM_001292028.1 Intron NP_001278957.1

View Full Product Details