Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001253792.1 | Intron | NP_001240721.1 | |||
NM_018337.3 | Intron | NP_060807.2 | |||
XM_005259034.2 | Intron | XP_005259091.1 | |||
XM_005259035.1 | Intron | XP_005259092.1 | |||
XM_005259036.1 | Intron | XP_005259093.1 | |||
XM_011527068.1 | Intron | XP_011525370.1 | |||
XM_011527069.1 | Intron | XP_011525371.1 | |||
XM_011527070.1 | Intron | XP_011525372.1 | |||
XM_011527071.1 | Intron | XP_011525373.1 | |||
XM_011527073.1 | Intron | XP_011525375.1 | |||
XM_017026932.1 | Intron | XP_016882421.1 | |||
XM_017026933.1 | Intron | XP_016882422.1 |