Product Details

SNP ID
rs651600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76875635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTGAGACTCACTATCTTGTATG[C/T]GGTTGTAGTCACTTGCACTTTTTTT
Phenotype
MIM: 612441
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT5B PubMed Links

Gene Details

Gene
MGAT5B
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199172.1 Intron NP_001186101.1
NM_144677.2 Intron NP_653278.2
NM_198955.1 Intron NP_945193.1
XM_006721707.3 Intron XP_006721770.1
XM_011524350.2 Intron XP_011522652.1
XM_011524352.2 Intron XP_011522654.1
XM_011524354.2 Intron XP_011522656.1
XM_017024200.1 Intron XP_016879689.1

View Full Product Details