Product Details

SNP ID
rs505481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143501311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTTCGGTTCCTATTTGATCAG[A/G]CAGTACTCAGGTGGCAACTGAAAGC
Phenotype
MIM: 136820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUCA2 PubMed Links

Gene Details

Gene
FUCA2
Gene Name
fucosidase, alpha-L- 2, plasma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032020.4 Intron NP_114409.2

View Full Product Details