Product Details

SNP ID
rs387834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50558455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTTATGGAGAGGCCAAGTGCA[C/G]GGACCCATCTCCCCACTCTCTAAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C3orf18 PubMed Links

Gene Details

Gene
C3orf18
Gene Name
chromosome 3 open reading frame 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171740.2 2260 UTR 3 NP_001165211.1
NM_001171741.2 2260 UTR 3 NP_001165212.1
NM_001171743.2 2260 UTR 3 NP_001165214.2
NM_016210.4 2260 UTR 3 NP_057294.2
XM_011533781.2 2260 UTR 3 XP_011532083.1
XM_011533782.2 2260 UTR 3 XP_011532084.1
XM_011533783.2 2260 UTR 3 XP_011532085.1
XM_011533784.2 2260 UTR 3 XP_011532086.1
XM_011533785.2 2260 UTR 3 XP_011532087.1
XM_011533788.1 2260 UTR 3 XP_011532090.1
XM_011533789.1 2260 UTR 3 XP_011532091.1
XM_011533790.1 2260 UTR 3 XP_011532092.1
XM_017006545.1 2260 UTR 3 XP_016862034.1
XM_017006546.1 2260 UTR 3 XP_016862035.1
XM_017006547.1 2260 UTR 3 XP_016862036.1

View Full Product Details