Product Details

SNP ID
rs501957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31987485 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCAGTGTTGTAAAATCAAAGAAG[A/G]GAAGCACTGGCAGTCATTGTCAGTG
Phenotype
MIM: 606245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUZ12 PubMed Links

Gene Details

Gene
SUZ12
Gene Name
SUZ12 polycomb repressive complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321207.1 Intron NP_001308136.1
NM_015355.3 Intron NP_056170.2
XM_006721794.3 Intron XP_006721857.1
XM_017024409.1 Intron XP_016879898.1

View Full Product Details