Product Details
- SNP ID
-
rs345187
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:60156155 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGTTTCCAAAGCCCTCTCCATCC[A/G]CCTCCTTTAAACTCTCTGACATCCC
- Phenotype
-
MIM: 114760
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CA4
PubMed Links
Gene Details
- Gene
- CA4
- Gene Name
- carbonic anhydrase 4
View Full Product Details