Product Details
- SNP ID
-
rs193205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:60150823 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCGCGGCCAGAGCAAGGGCGGT[C/G]TGGCAGGGTCCTGGAAGCCGCGCTT
- Phenotype
-
MIM: 114760
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CA4
PubMed Links
Gene Details
- Gene
- CA4
- Gene Name
- carbonic anhydrase 4
View Full Product Details