Product Details

SNP ID
rs511819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63698212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTCAGCTCACTGCCACCTCCGC[C/T]TCCTGGGCTCAAGTGATCCTTTCAG
Phenotype
MIM: 604249
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTN3 PubMed Links

Gene Details

Gene
RTN3
Gene Name
reticulon 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265589.1 Intron NP_001252518.1
NM_001265590.1 Intron NP_001252519.1
NM_001265591.1 Intron NP_001252520.1
NM_006054.3 Intron NP_006045.1
NM_201428.2 Intron NP_958831.1
NM_201429.2 Intron NP_958832.1
NM_201430.2 Intron NP_958833.1
XM_011544730.2 Intron XP_011543032.1
XM_011544731.2 Intron XP_011543033.1
XM_017017091.1 Intron XP_016872580.1
XM_017017092.1 Intron XP_016872581.1

View Full Product Details