Product Details

SNP ID
rs688568
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32192114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTATTATTGTTTTTGTTATTT[G/T]GTGTCAAATCAGATGGACCTAATAA
Phenotype
MIM: 600389
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MEP1B PubMed Links

Gene Details

Gene
MEP1B
Gene Name
meprin A subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308171.1 Intron NP_001295100.1
NM_005925.2 Intron NP_005916.2
XM_011526013.2 Intron XP_011524315.1
XM_011526014.2 Intron XP_011524316.1

View Full Product Details