Product Details

SNP ID
rs368333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3136969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCGTTCTGGCCAGCCCACCCG[C/T]AAGGGAGGGGCCGGCTCCAGCCTCT
Phenotype
MIM: 139314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNA15 PubMed Links

Gene Details

Gene
GNA15
Gene Name
G protein subunit alpha 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002068.3 Intron NP_002059.3
Gene
LOC100996351
Gene Name
uncharacterized LOC100996351
There are no transcripts associated with this gene.

View Full Product Details