Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243723.1 | Intron | NP_001230652.1 | |||
NM_001276325.1 | Intron | NP_001263254.1 | |||
NM_001320676.1 | Intron | NP_001307605.1 | |||
NM_001320677.1 | Intron | NP_001307606.1 | |||
NM_001320678.1 | Intron | NP_001307607.1 | |||
NM_006515.3 | Intron | NP_006506.3 | |||
XM_006713292.3 | Intron | XP_006713355.1 | |||
XM_006713294.3 | Intron | XP_006713357.1 | |||
XM_017007032.1 | Intron | XP_016862521.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164674.1 | Intron | NP_001158146.1 | |||
NM_001164675.1 | Intron | NP_001158147.1 | |||
NM_182760.3 | Intron | NP_877437.2 | |||
XM_011533624.2 | Intron | XP_011531926.1 | |||
XM_011533625.2 | Intron | XP_011531927.1 | |||
XM_011533626.2 | Intron | XP_011531928.1 | |||
XM_017006252.1 | Intron | XP_016861741.1 | |||
XM_017006253.1 | Intron | XP_016861742.1 | |||
XM_017006254.1 | Intron | XP_016861743.1 | |||
XM_017006255.1 | Intron | XP_016861744.1 |