Product Details

SNP ID
rs613686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78171347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGTTATGTATTACTGTTATGCC[C/T]ATTTCACAGATGAAGAAATGGAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD21 PubMed Links

Gene Details

Gene
KCTD21
Gene Name
potassium channel tetramerization domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029859.2 3294 UTR 3 NP_001025030.1
XM_005273925.3 3294 UTR 3 XP_005273982.1
XM_005273928.1 3294 UTR 3 XP_005273985.1
XM_006718516.3 3294 UTR 3 XP_006718579.1
XM_006718517.2 3294 UTR 3 XP_006718580.1
XM_006718518.3 3294 UTR 3 XP_006718581.1
XM_011544955.2 3294 UTR 3 XP_011543257.1
Gene
KCTD21-AS1
Gene Name
KCTD21 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details