Product Details

SNP ID
rs293602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:59748646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGACTATTTGGAGGGTACAAGG[A/G]GAGATGGGACAGGGAGAAGGAAGTG
Phenotype
MIM: 601153
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FHIT PubMed Links

Gene Details

Gene
FHIT
Gene Name
fragile histidine triad
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166243.2 4321 UTR 3 NP_001159715.1
NM_001320899.1 4321 UTR 3 NP_001307828.1
NM_001320900.1 4321 UTR 3 NP_001307829.1
NM_001320901.1 4321 UTR 3 NP_001307830.1
NM_002012.3 4321 UTR 3 NP_002003.1
XM_017005880.1 4321 Intron XP_016861369.1
XM_017005881.1 4321 Intron XP_016861370.1
XM_017005882.1 4321 Intron XP_016861371.1
XM_017005883.1 4321 Intron XP_016861372.1
XM_017005884.1 4321 Intron XP_016861373.1
XM_017005885.1 4321 Intron XP_016861374.1
XM_017005886.1 4321 Intron XP_016861375.1

View Full Product Details