Product Details

SNP ID
rs3160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2209190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGTGCCCACCCCCAAGCTAGTG[C/T]GTTCTCTGCCCCTCCCTGCCCGCGT
Phenotype
MIM: 612190 MIM: 172280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRICD5 PubMed Links

Gene Details

Gene
BRICD5
Gene Name
BRICHOS domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182563.3 1650 Intron NP_872369.2
XM_005255263.4 1650 Intron XP_005255320.1
Gene
MLST8
Gene Name
MTOR associated protein, LST8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199173.1 1650 UTR 3 NP_001186102.1
NM_001199174.1 1650 UTR 3 NP_001186103.1
NM_001199175.1 1650 UTR 3 NP_001186104.1
NM_022372.4 1650 UTR 3 NP_071767.3
XM_005255475.3 1650 UTR 3 XP_005255532.2
XM_005255478.2 1650 UTR 3 XP_005255535.1
XM_005255479.2 1650 UTR 3 XP_005255536.1
XM_017023548.1 1650 UTR 3 XP_016879037.1
XM_017023549.1 1650 UTR 3 XP_016879038.1
XM_017023550.1 1650 Intron XP_016879039.1
XM_017023551.1 1650 UTR 3 XP_016879040.1
XM_017023552.1 1650 Intron XP_016879041.1
Gene
PGP
Gene Name
phosphoglycolate phosphatase
There are no transcripts associated with this gene.

View Full Product Details