Product Details

SNP ID
rs627275
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95092368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGCCATTTATGCAGCTAGGGA[A/G]GGAGTGAGTCGTAAAGGAGCATAGT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ENDOD1 PubMed Links

Gene Details

Gene
ENDOD1
Gene Name
endonuclease domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015036.2 Intron NP_055851.1

View Full Product Details