Product Details
- SNP ID
-
rs244897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:168491787 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTTGCATACTCAAGTATTAACTT[C/T]TGTTATTGTCATTATTTTATACTGT
- Phenotype
-
MIM: 107820
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RARS
PubMed Links
Gene Details
- Gene
- RARS
- Gene Name
- arginyl-tRNA synthetase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002887.3 |
|
Intron |
|
|
NP_002878.2 |
View Full Product Details