Product Details

SNP ID
rs313842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46721060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCAACTCTCCCTGCCCCGCAGCA[C/T]TGACGCTGCAGCTGGGCAACAGGAA
Phenotype
MIM: 607074 MIM: 614767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987269 PubMed Links

Gene Details

Gene
LOC107987269
Gene Name
uncharacterized LOC107987269
There are no transcripts associated with this gene.

Gene
PRKD2
Gene Name
protein kinase D2
There are no transcripts associated with this gene.

Gene
STRN4
Gene Name
striatin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039877.1 Intron NP_001034966.1
NM_013403.2 Intron NP_037535.2
XM_006723171.2 Intron XP_006723234.1
XM_006723172.2 Intron XP_006723235.1
XM_006723174.2 Intron XP_006723237.1
XM_011526878.1 Intron XP_011525180.1
XM_011526879.1 Intron XP_011525181.1
XM_017026716.1 Intron XP_016882205.1
XM_017026717.1 Intron XP_016882206.1
XM_017026718.1 Intron XP_016882207.1
XM_017026719.1 Intron XP_016882208.1

View Full Product Details