Product Details

SNP ID
rs436187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47065264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGCAACACACCTTGGAACAC[C/G]TGCATCTGCTGGCTTGGGAAAGGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZC3H4 PubMed Links

Gene Details

Gene
ZC3H4
Gene Name
zinc finger CCCH-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015168.1 4823 UTR 3 NP_055983.1
XM_005258676.3 4823 Intron XP_005258733.1
XM_005258677.4 4823 Intron XP_005258734.1
XM_005258678.2 4823 Intron XP_005258735.1
XM_006723113.3 4823 Intron XP_006723176.1
XM_011526668.2 4823 Intron XP_011524970.1
XM_011526669.2 4823 Intron XP_011524971.1
XM_011526670.2 4823 Intron XP_011524972.1
XM_017026529.1 4823 Intron XP_016882018.1
XM_017026530.1 4823 UTR 3 XP_016882019.1
XM_017026531.1 4823 UTR 3 XP_016882020.1

View Full Product Details