Product Details

SNP ID
rs504003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55654178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCAGCGGTCGCGCGCACCCTTCA[C/T]CCCTCTTGCTGCTTTTGTCTGGCAC
Phenotype
MIM: 613478 MIM: 191318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC106 PubMed Links

Gene Details

Gene
CCDC106
Gene Name
coiled-coil domain containing 106
There are no transcripts associated with this gene.

Gene
U2AF2
Gene Name
U2 small nuclear RNA auxiliary factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012478.1 47 UTR 5 NP_001012496.1
NM_007279.2 47 UTR 5 NP_009210.1
XM_006722994.2 47 UTR 5 XP_006723057.1
XM_011526410.1 47 UTR 5 XP_011524712.1
Gene
ZNF581
Gene Name
zinc finger protein 581
There are no transcripts associated with this gene.

View Full Product Details