Product Details

SNP ID
rs491639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55544818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACCCCCCTGGTGAGGCTGGGC[A/G]AGGAGCACGCGGCCGTAGGAGCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
There are no transcripts associated with this gene.

Gene
SBK3
Gene Name
SH3 domain binding kinase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199824.2 177 Silent Mutation CTC,CTT L59L NP_001186753.1
XM_011526298.2 177 Silent Mutation CTC,CTT L84L XP_011524600.1

View Full Product Details