Product Details

SNP ID
rs291989
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:22653406 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTTGCGTCACCTGCCTCGGAGCCAG[C/T]TAGGCTGTTTAACAGTGCAGGTAGC
Phenotype
MIM: 120570 MIM: 120575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QB PubMed Links

Gene Details

Gene
C1QB
Gene Name
complement C1q B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000491.3 Intron NP_000482.3
XM_011542059.2 Intron XP_011540361.1
Gene
C1QC
Gene Name
complement C1q C chain
There are no transcripts associated with this gene.

View Full Product Details